Orders LinkContactUsLinkComponent
Dropping funnels, cylindrical, without pressure balance
Supplier:   Lenz Laborglas
Dropping funnels, cylindrical, without pressure balance
Supplier:  Lenz Laborglas
Certificates
Dropping funnels, cylindrical, without pressure balance
Dropping funnels, cylindrical, without pressure balance
8 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear, with PTFE key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible white divisions for approximate volume reading.
DIN ISO 4800
 : DIN ISO 4800
Product Family Options
Product Information
  • Capacity
    Cone size
    Socket size
    Division
    Availability
    Price
  • 50 ml
    29/32 NS
    29/32 NS
    1 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 88,40 €
    88,40 €
    1 * 1 ST
       88,40 €
  • 250 ml
    29/32 NS
    29/32 NS
    5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
  • 500 ml
    29/32 NS
    29/32 NS
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 121,00 €
    121,00 €
    1 * 1 ST
       121,00 €
  • 1000 ml
    29/32 NS
    29/32 NS
    20 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 153,00 €
    153,00 €
    1 * 1 ST
       153,00 €
  • 50 ml
    14/23 NS
    14/23 NS
    1 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 91,00 €
    91,00 €
    1 * 1 ST
       91,00 €
  • 100 ml
    14/23 NS
    14/23 NS
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 85,90 €
    85,90 €
    1 * 1 ST
       85,90 €
  • 100 ml
    29/32 NS
    29/32 NS
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,50 €
    94,50 €
    1 * 1 ST
       94,50 €
  • 25 ml
    14/23 NS
    14/23 NS
    1 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,50 €
    80,50 €
    1 * 1 ST
       80,50 €