Orders LinkContactUsLinkComponent
Funnels with short stem
Funnels with short stem
Supplier:   KARTELL
Funnels with short stem
Supplier:  KARTELL
Certificates
Funnels with short stem
Funnels with short stem
9 Options Available Below
Product Details & Documents
HDPE, translucent.
  • Good chemical resistance
  • With raised lip to prevent overflowing
  • External ribs for unobstructed removal of air when filtering
With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
Documents
Product Family Options
Product Information
  • Length
    Funnel O-Ø
    Availability
    Price
  • 80 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,80 €
    3,80 €
    1 * 1 ST
       3,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    221-1263
  • 100 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,00 €
    4,00 €
    1 * 1 ST
       4,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    221-1521
  • 126 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,60 €
    4,60 €
    1 * 1 ST
       4,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    221-1522
  • 154 mm
    140 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,10 €
    6,10 €
    1 * 1 ST
       6,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    221-1523
  • 180 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,00 €
    10,00 €
    1 * 1 ST
       10,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    221-1524
  • 230 mm
    220 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,30 €
    12,30 €
    1 * 1 ST
       12,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    221-1525
  • 240 mm
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,30 €
    17,30 €
    1 * 1 ST
       17,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    221-0080
  • 280 mm
    310 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,20 €
    22,20 €
    1 * 1 ST
       22,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    221-0081
  • 360 mm
    420 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,80 €
    38,80 €
    1 * 1 ST
       38,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    221-0082