Order Entry
Belgium
Orders LinkContactUsLinkComponent
Funnels, powder

Funnels, powder

80 mm, 25 mm, 25 mm
Catalog # 221-0002
Supplier:   SciLabware
Funnels, powder
Catalog # 221-0002
Supplier:  SciLabware
Funnels, powder
Funnels, powder

80 mm, 25 mm, 25 mm

Price
333,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

PYREX®, borosilicate glass.

  • Chemically resistant glass
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Quick and easy to clean

Ideal for transferring powders, can also be used for liquids.

Specifications
  • Funnel O-Ø:
    80 mm
  • Stem O-Ø:
    25 mm
  • Stem length:
    25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.