Orders LinkContactUsLinkComponent
Funnels, powder
Supplier:   Kavalier
Funnels, powder
Supplier:  Kavalier
Certificates
Funnels, powder
7 Options Available Below
Product Details & Documents
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With smooth internal walls. For transferring powdery materials. Also suitable for use with liquids. Funnel rim is ground and fire-polished.
Product Family Options
Product Information
  • Funnel O-Ø
    Stem O-Ø
    Stem length
    Availability
    Price
  • 50 mm
    20 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,10 €
    29,10 €
    1 * 1 ST
       29,10 €
  • 60 mm
    20 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,60 €
    29,60 €
    1 * 1 ST
       29,60 €
  • 80 mm
    25 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,40 €
    34,40 €
    1 * 1 ST
       34,40 €
  • 100 mm
    25 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 3 090,00 €
    3 090,00 €
    1 * 10 ST
       3 090,00 €
  • 120 mm
    30 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,00 €
    46,00 €
    1 * 1 ST
       46,00 €
  • 160 mm
    35 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,70 €
    65,70 €
    1 * 1 ST
       65,70 €
  • 200 mm
    38 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 110,00 €
    110,00 €
    1 * 1 ST
       110,00 €