Orders LinkContactUsLinkComponent
Funnels with conical joint
Funnels with conical joint
Supplier:   Lenz Laborglas
Funnels with conical joint
Supplier:  Lenz Laborglas
Certificates
Funnels with conical joint
Funnels with conical joint
4 Options Available Below
Product Details & Documents
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.
Documents
Product Family Options
Product Information
  • Cone size
    Funnel O-Ø
    Availability
    Price
  • 24/29 NS
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,10 €
    40,10 €
    1 * 1 ST
       40,10 €
  • 14/23 NS
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,50 €
    21,50 €
    1 * 1 ST
       21,50 €
  • 29/32 NS
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,40 €
    40,40 €
    1 * 1 ST
       40,40 €
  • 19/26 NS
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,60 €
    24,60 €
    1 * 1 ST
       24,60 €