Orders LinkContactUsLinkComponent
Funnels, drum
Funnels, drum
Supplier:   VITLAB
Funnels, drum
Supplier:  VITLAB
Certificates
Funnels, drum
5 Options Available Below
Product Details & Documents
PP or HDPE, translucent.
  • Very good (PP) or good (HDPE) chemical resistance
  • Filter insert made from stainless steel and aluminium is available separately for 200 and 250 mm sizes
  • Quick and easy to clean
For filling containers with large quantities of liquid or powdered substances quickly.
Product Family Options
Product Information
  • Funnel O-Ø
    Material
    Stem O-Ø
    Length
    Availability
    Price
  • 200 mm
    PP
    22 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,40 €
    11,40 €
    1 * 1 ST
       11,40 €
  • 250 mm
    PP
    30 mm
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,40 €
    19,40 €
    1 * 1 ST
       19,40 €
  • 350 mm
    PP
    35 mm
    440 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,80 €
    40,80 €
    1 * 1 ST
       40,80 €
  • 430 mm
    HDPE
    37 mm
    420 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 132,00 €
    132,00 €
    1 * 1 ST
       132,00 €
  • 400 mm
    HDPE
    42 mm
    365 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,40 €
    98,40 €
    1 * 1 ST
       98,40 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Sieve insert, Ø 50 mm, for funnels 221-1411 and 221-1412
    Sieve insert, Ø 50 mm, for funnels 221-1411 and 221-1412
    221-1416
    Log in for availability
    1 * 1 ST - 6,70 €