Orders LinkContactUsLinkComponent
Analytical funnel
Analytical funnel
Supplier:   Kavalier
Analytical funnel
Supplier:  Kavalier
Certificates
Analytical funnel
Analytical funnel
3 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For rapid filtration.
Product Family Options
Product Information
  • Stem length
    Stem O-Ø
    Funnel O-Ø
    Availability
    Price
  • 150 mm
    8 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,40 €
    45,40 €
    1 * 1 ST
       45,40 €
  • 150 mm
    8 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,20 €
    51,20 €
    1 * 1 ST
       51,20 €
  • 150 mm
    8 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,30 €
    58,30 €
    1 * 1 ST
       58,30 €