Orders LinkContactUsLinkComponent
Funnels
Funnels
Supplier:   NICKEL ELECTRO
Funnels
Supplier:  NICKEL ELECTRO
Certificates
Funnels
3 Options Available Below
Product Details & Documents
Stainless steel.
  • Good chemical resistance, extremely high thermal resistance
  • With air duct for discharging container air
  • Autoclavable
Smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
Product Family Options
Product Information
  • Funnel O-Ø
    Availability
    Price
  • 100 m mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,60 €
    24,60 €
    1 * 1 ST
       24,60 €
  • 150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,10 €
    37,10 €
    1 * 1 ST
       37,10 €
  • 200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,40 €
    51,40 €
    1 * 1 ST
       51,40 €