Orders LinkContactUsLinkComponent
Forceps
Forceps
Supplier:   KARTELL
Forceps
Supplier:  KARTELL
Certificates
Forceps
3 Options Available Below
Product Details & Documents
Polyoxymethylene (POM), yellow. Multipurpose forceps for a wide range of applications.
  • Good chemical resistance, not suitable for use with strong acids and solvents
  • Autoclavable
  • Robust, pliable, self-sprung
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Blunt
    118 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 32,70 €
    32,70 €
    1 * 5 ST
       32,70 €
  • Blunt
    147 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 41,60 €
    41,60 €
    1 * 5 ST
       41,60 €
  • Blunt
    253 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 124,00 €
    124,00 €
    1 * 5 ST
       124,00 €