Orders LinkContactUsLinkComponent
Forceps with guide pin
Forceps with guide pin
Supplier:   BOCHEM
Forceps with guide pin
Supplier:  BOCHEM
Certificates
Forceps with guide pin
Forceps with guide pin
3 Options Available Below
Product Details & Documents
Stainless steel, PTFE coating.
  • Anti-magnetic
  • Very good chemical and thermal resistance
  • Smooth surface prevents contamination and facilitates cleaning
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Sharp, straight
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,10 €
    35,10 €
    1 * 1 ST
       35,10 €
  • Sharp, straight
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,10 €
    35,10 €
    1 * 1 ST
       35,10 €
  • Sharp, straight
    145 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €