Orders LinkContactUsLinkComponent
Forceps, precision, with locking clip
Forceps, precision, with locking clip
Forceps, precision, with locking clip
Supplier:  OUTILS RUBIS
Certificates
Forceps, precision, with locking clip
Forceps, precision, with locking clip
2 Options Available Below
Product Details & Documents
Stainless steel. Forceps with very precise, fine tips. For applications that require high precision such as in microscopy, biology and microbiology.
  • Acid-resistant, resistant to hydrofluoric acid and nitric acid
  • 100% antimagnetic
  • Extra fine tips
Documents
Certificates
Search by Lot Number
    Images 360
  • 5L
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • 232-1521
    Straight, ultra fine tips
    Straight, ultra fine tips
    3L, with locking clip
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,80 €
    51,80 €
    1 * 1 ST
       51,80 €
  • 232-1522
    Straight, ultra fine tips (tower tip)
    Straight, ultra fine tips (tower tip)
    5L, with locking clip
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,00 €
    45,00 €
    1 * 1 ST
       45,00 €