Orders LinkContactUsLinkComponent
Forceps with guide pin
Forceps with guide pin
Supplier:   RSG
Forceps with guide pin
Supplier:  RSG
Certificates
Forceps with guide pin
Forceps with guide pin
6 Options Available Below
Product Details & Documents
Stainless steel, anti-magnetic. Particularly suitable for staining applications.
  • With curved, extra fine tips
  • Corrosion resistant
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Curved, with guide pin
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,60 €
    11,60 €
    1 * 1 ST
       11,60 €
  • Curved, with guide pin
    145 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,80 €
    11,80 €
    1 * 1 ST
       11,80 €
  • Curved, with guide pin
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,40 €
    14,40 €
    1 * 1 ST
       14,40 €
  • Curved, with guide pin
    200 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 101,00 €
    101,00 €
    1 * 5 ST
       101,00 €
  • Curved, with guide pin
    115 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 56,50 €
    56,50 €
    1 * 5 ST
       56,50 €
  • Curved, with guide pin
    105 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 54,90 €
    54,90 €
    1 * 5 ST
       54,90 €