Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Supplier:   BOCHEM
Forceps, precision
Supplier:  BOCHEM
Certificates
Forceps, precision
Forceps, precision
3 Options Available Below
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Extremely precise forceps designed for a wide variety of uses in the laboratory, production and more.
  • Extremely sharp, flat
  • Non serrated points
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Straight
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,00 €
    9,00 €
    1 * 1 ST
       9,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    232-0007
  • Curved
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,00 €
    10,00 €
    1 * 1 ST
       10,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    232-0008
  • Straight
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,70 €
    13,70 €
    1 * 1 ST
       13,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    BOCH1611