Orders LinkContactUsLinkComponent
Forceps with guide pin
 :   BOCHEM
Forceps with guide pin
 :  BOCHEM
Forceps with guide pin
Forceps with guide pin
Stainless steel, with PTFE coating.
  • Bent, with guide pin
  • Smooth, coated surface to prevent contamination and facilitate cleaning
Product Family Options
  • Type
    Length
  • Bent, sharp
    115 mm
    1 * 1 ST
    1 * 1 ST - 35,70 €
    35,70 €
    1 * 1 ST
       35,70 €
  • Bent, sharp
    130 mm
    1 * 1 ST
    1 * 1 ST - 35,70 €
    35,70 €
    1 * 1 ST
       35,70 €
  • Bent, sharp
    145 mm
    1 * 1 ST
    1 * 1 ST - 36,70 €
    36,70 €
    1 * 1 ST
       36,70 €