Forceps, precision
Supplier: OUTILS RUBIS
Jump to:
About this item
Stainless steel. Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology and electron microscopy.
- Acid-resistant
- Resistant against hydrogen fluoride and nitric acid
- 100% anti-magnetic
16 Options Available Below
Product Details & Documents
Stainless steel. Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology and electron microscopy.
- Acid-resistant
- Resistant against hydrogen fluoride and nitric acid
- 100% anti-magnetic
Documents
Product Family Options
Product Information
- TypeLengthAvailabilityPrice
- 232-1132Serrated handle, straight, pointed, ultra fine tips
- 232-1219Straight, extra strong fine tips
- 232-1220Straight, short, extra fine tips
- 232-1221Straight, short fine tips
- 232-1230Curved, extra fine tips
- OUTI5BBent, needle pointed, ultra fine tips
- 232-0000Straight, short fine tips
- 232-0143Straight, extra fine tips
- 232-0216Straight, short fine tips
- 232-0209Straight, extra wide flat tips
- OUTI35BSerrated handle, curved, round, broad, flat tips
- 232-0217Straight, very fine tips
- OUTIFStraight, squared, flat, fine tips
- OUTI6Angled, pointed, fine tips
- OUTI0Straight, pointed, ultra fine tips
- OUTISS-SAStraight, pointed, ultra fine tips