Volumetric flasks, class A, blue markings
MBL®, borosilicate glass, with PE stopper.
- Marks and inscriptions in high contrast blue enamel, conformity 'H' mark
- Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
- Calibrated to contain (TC, In)
: DIN 12664, BS EN ISO 1042
Frequently Bought Together