Volumetric flasks, class A, white markings
Supplier: Poulten & Graf
Certificates
About this item
Borosilicate glass, amber, with PP NS-stopper.
- Easy to read white markings
- Very good chemical resistance
- Very good temperature resistance
Amber coloured to protect light sensitive substances.
Certifications: Acc. to ISO and ASTM or USP standards
Frequently Bought Together