Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
1000 ml, 24/29, 0,40 ± ml
Catalog # 612-3807
Supplier:   VITLAB
Volumetric flasks, class A
Catalog # 612-3807
Supplier:  VITLAB
Volumetric flasks, class A
Volumetric flasks, class A
1000 ml, 24/29, 0,40 ± ml
Price
38,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
PMP (TPX®), transparent, with PP stopper.
  • Individually calibrated ring mark
  • Imprinted lot number
  • Complete with certificate
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    24/29
  • Capacity:
    1000 ml
  • Seq:
    06
  • Tolerance:
    0,40 ± ml