Order Entry
Belgium
ContactUsLinkComponent
 
Volumetric flasks, class B, red markings
  612-3916
 :  VITLAB
undefined
Volumetric flasks, class B, red markings
  612-3916
 :  VITLAB

 

  • NS:
    24/29
  • Capacity:
    1000 ml
  • Pk:
    1
  • Tolerance:
    0,60 ± ml

 

 

PMP, translucent, with PP NS-stopper.

  • Tolerances are approximately 20% better than class B according to DIN EN ISO 1042
  • Individually ring marked
  • With NS stoppers
 : DIN EN ISO 1042