Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
Supplier:   KARTELL
Volumetric flasks, class B
Supplier:  KARTELL
Certificates
Volumetric flasks, class B
Volumetric flasks, class B
6 Options Available Below
Product Details & Documents
PP, translucent, with PE stopper.
  • Narrow neck with no meniscus for easy reading
  • Calibrated gravimetrically at +20 °C
  • Autoclavable, chemically non absorbent, non wetting surface
Product Family Options
Product Information
  • Capacity
    NS
    Height
    Availability
    Price
  • 25 ml
    12/21
    121 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,10 €
    17,10 €
    1 * 1 ST
       17,10 €
  • 50 ml
    14/23
    143 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,00 €
    18,00 €
    1 * 1 ST
       18,00 €
  • 100 ml
    14/23
    174 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,50 €
    21,50 €
    1 * 1 ST
       21,50 €
  • 250 ml
    19/26
    225 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,10 €
    24,10 €
    1 * 1 ST
       24,10 €
  • 500 ml
    19/26
    265 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,50 €
    27,50 €
    1 * 1 ST
       27,50 €
  • 1000 ml
    19/26
    325 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,30 €
    34,30 €
    1 * 1 ST
       34,30 €