Orders LinkContactUsLinkComponent
Gas sample tubes
Gas sample tubes
Supplier:   Lenz Laborglas
Gas sample tubes
Supplier:  Lenz Laborglas
Certificates
Gas sample tubes
Gas sample tubes
4 Options Available Below
Product Details & Documents
Duran®, borosilicate glass, clear. Both ends with a 3 NS/14 one-way stop cock. Sampling tube with GL 14 thread.
  • White labelling area
  • Excellent thermal and chemical resistance
  • Connection on one end with capillary tube (i. Ø: 2,7 mm ; o. Ø: 8,0 mm) and on the other end with tube connector (o. Ø: 8,0 mm)
Gas collection tubes for removing, storing and transporting gas samples.
Certifications: DIN 12473-1
Documents
Product Family Options
Product Information
  • Capacity
    Type
    Availability
    Price
  • 150 ml
    with nozzle (GL 14) and septa
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 111,00 €
    111,00 €
    1 * 1 ST
       111,00 €
  • 350 ml
    with nozzle (GL 14) and septa
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 122,00 €
    122,00 €
    1 * 1 ST
       122,00 €
  • 500 ml
    with nozzle (GL 14) and septa
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 134,00 €
    134,00 €
    1 * 1 ST
       134,00 €
  • 1000 ml
    with nozzle (GL 14) and septa
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 170,00 €
    170,00 €
    1 * 1 ST
       170,00 €