Orders LinkContactUsLinkComponent
Gas sampling tubes
Supplier:   Lenz Laborglas
Gas sampling tubes
Supplier:  Lenz Laborglas
Certificates
Gas sampling tubes
Gas sampling tubes
4 Options Available Below
Product Details & Documents
Duran® clear borosilicate glass.
  • Excellent thermal and chemical resistance
  • Connection on one side with capillary tube (internal Ø: 2,7 mm, external Ø: 8,0 mm) and on the other side with tube connector (external Ø: 8,0 mm)
  • With white labelling area
Gas collection tubes for removing, collecting and transporting gas samples. Both sides with a 3 NS/14 standard ground one-way stop cock.
 : DIN 12473-1
Product Family Options
Product Information
  • Capacity
    Type
    Availability
    Price
  • 500 ml
    without threaded bleed connection
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 118,00 €
    118,00 €
    1 * 1 ST
       118,00 €
  • 1000 ml
    without threaded bleed connection
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 151,00 €
    151,00 €
    1 * 1 ST
       151,00 €
  • 150 ml
    without threaded bleed connection
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,50 €
    93,50 €
    1 * 1 ST
       93,50 €
  • 350 ml
    without threaded bleed connection
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €