Orders LinkContactUsLinkComponent
Flasks, flat bottom, narrow neck
Flasks, flat bottom, narrow neck
Supplier:   SciLabware
Flasks, flat bottom, narrow neck
Supplier:  SciLabware
Certificates
Flasks, flat bottom, narrow neck
Flasks, flat bottom, narrow neck
8 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, clear.
  • High resistance to thermal shock
  • Excellent chemical durability for contamination-free performance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Narrow neck flasks with flat base for standing on benches and hotplates. Marked with capacity and a white labelling area.
Certifications: ISO 1773
Product Family Options
Product Information
  • Capacity
    Height
    Neck I-Ø
    Ext.Ø
    Availability
    Price
  • 100 ml
    110 mm
    18 mm
    64 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 98,50 €
    98,50 €
    1 * 10 ST
       98,50 €
  • 250 ml
    140 mm
    30 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 116,00 €
    116,00 €
    1 * 10 ST
       116,00 €
  • 500 ml
    170 mm
    30 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,70 €
    15,70 €
    1 * 1 ST
       15,70 €
  • 1000 ml
    200 mm
    36 mm
    131 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,00 €
    23,00 €
    1 * 1 ST
       23,00 €
  • 2000 ml
    250 mm
    45 mm
    166 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,20 €
    42,20 €
    1 * 1 ST
       42,20 €
  • 4000 ml
    300 mm
    45 mm
    207 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 6000 ml
    340 mm
    57 mm
    237 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 152,00 €
    152,00 €
    1 * 1 ST
       152,00 €
  • 10000 ml
    400 mm
    57 mm
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 264,00 €
    264,00 €
    1 * 1 ST
       264,00 €