Orders LinkContactUsLinkComponent
Flasks, round bottom, Quickfit® ground joints
 :   SciLabware
Flasks, round bottom, Quickfit® ground joints
Flasks, round bottom, Quickfit® ground joints
Flasks, round bottom, Quickfit® ground joints
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
 : ISO 1773, ISO 4797.
Product Family Options
  • Capacity
    Socket size
    Height
  • 5 ml
    10/19 NS
    50 mm
    1 * 20 ST
    1 * 20 ST - 252,00 €
    252,00 €
    1 * 20 ST
       252,00 €
  • 5 ml
    14/23 NS
    65 mm
    1 * 5 ST
    1 * 5 ST - 86,50 €
    86,50 €
    1 * 5 ST
       86,50 €
  • 10 ml
    10/19 NS
    68 mm
    1 * 5 ST
    1 * 5 ST - 70,20 €
    70,20 €
    1 * 5 ST
       70,20 €
  • 10 ml
    14/23 NS
    68 mm
    1 * 5 ST
    1 * 5 ST - 86,40 €
    86,40 €
    1 * 5 ST
       86,40 €
  • 25 ml
    14/23 NS
    80 mm
    1 * 5 ST
    1 * 5 ST - 46,30 €
    46,30 €
    1 * 5 ST
       46,30 €
  • 25 ml
    19/26 NS
    80 mm
    1 * 5 ST
    1 * 5 ST - 76,30 €
    76,30 €
    1 * 5 ST
       76,30 €
  • 25 ml
    24/29 NS
    80 mm
    1 * 5 ST
    1 * 5 ST - 76,40 €
    76,40 €
    1 * 5 ST
       76,40 €
  • 50 ml
    14/23 NS
    90 mm
    1 * 5 ST
    1 * 5 ST - 60,60 €
    60,60 €
    1 * 5 ST
       60,60 €
  • 50 ml
    19/26 NS
    90 mm
    1 * 5 ST
    1 * 5 ST - 103,00 €
    103,00 €
    1 * 5 ST
       103,00 €
  • 50 ml
    24/29 NS
    95 mm
    1 * 5 ST
    1 * 5 ST - 92,90 €
    92,90 €
    1 * 5 ST
       92,90 €
  • 100 ml
    14/23 NS
    108 mm
    1 * 5 ST
    1 * 5 ST - 65,40 €
    65,40 €
    1 * 5 ST
       65,40 €
  • 100 ml
    19/26 NS
    108 mm
    1 * 5 ST
    1 * 5 ST - 119,00 €
    119,00 €
    1 * 5 ST
       119,00 €
  • 100 ml
    24/29 NS
    110 mm
    1 * 5 ST
    1 * 5 ST - 110,00 €
    110,00 €
    1 * 5 ST
       110,00 €
  • 100 ml
    34/35 NS
    112 mm
    1 * 5 ST
    1 * 5 ST - 104,00 €
    104,00 €
    1 * 5 ST
       104,00 €
  • 100 ml
    29/32 NS
    114 mm
    1 * 5 ST
    1 * 5 ST - 119,00 €
    119,00 €
    1 * 5 ST
       119,00 €
  • 150 ml
    19/26 NS
    114 mm
    1 * 5 ST
    1 * 5 ST - 109,00 €
    109,00 €
    1 * 5 ST
       109,00 €
  • 150 ml
    24/29 NS
    115 mm
    1 * 5 ST
    1 * 5 ST - 99,60 €
    99,60 €
    1 * 5 ST
       99,60 €
  • 150 ml
    34/35 NS
    118 mm
    1 * 5 ST
    1 * 5 ST - 109,00 €
    109,00 €
    1 * 5 ST
       109,00 €
  • 150 ml
    29/32 NS
    120 mm
    1 * 5 ST
    1 * 5 ST - 98,90 €
    98,90 €
    1 * 5 ST
       98,90 €
  • 250 ml
    14/23 NS
    126 mm
    1 * 2 ST
    1 * 2 ST - 49,80 €
    49,80 €
    1 * 2 ST
       49,80 €
  • 250 ml
    19/26 NS
    129 mm
    1 * 5 ST
    1 * 5 ST - 107,00 €
    107,00 €
    1 * 5 ST
       107,00 €
  • 250 ml
    24/29 NS
    130 mm
    1 * 5 ST
    1 * 5 ST - 105,00 €
    105,00 €
    1 * 5 ST
       105,00 €
  • 250 ml
    29/32 NS
    135 mm
    1 * 5 ST
    1 * 5 ST - 84,60 €
    84,60 €
    1 * 5 ST
       84,60 €
  • 250 ml
    34/35 NS
    138 mm
    1 * 5 ST
    1 * 5 ST - 112,00 €
    112,00 €
    1 * 5 ST
       112,00 €
  • 500 ml
    19/26 NS
    147 mm
    1 * 5 ST
    1 * 5 ST - 97,30 €
    97,30 €
    1 * 5 ST
       97,30 €
  • 500 ml
    24/29 NS
    150 mm
    1 * 5 ST
    1 * 5 ST - 114,00 €
    114,00 €
    1 * 5 ST
       114,00 €
  • 500 ml
    29/32 NS
    153 mm
    1 * 5 ST
    1 * 5 ST - 118,00 €
    118,00 €
    1 * 5 ST
       118,00 €
  • 500 ml
    34/35 NS
    166 mm
    1 * 5 ST
    1 * 5 ST - 139,00 €
    139,00 €
    1 * 5 ST
       139,00 €
  • 1000 ml
    19/26 NS
    177 mm
    1 * 10 ST
    1 * 10 ST - 351,00 €
    351,00 €
    1 * 10 ST
       351,00 €
  • 1000 ml
    24/29 NS
    180 mm
    1 * 2 ST
    1 * 2 ST - 53,20 €
    53,20 €
    1 * 2 ST
       53,20 €
  • 1000 ml
    29/32 NS
    183 mm
    1 * 2 ST
    1 * 2 ST - 58,00 €
    58,00 €
    1 * 2 ST
       58,00 €
  • 1000 ml
    34/35 NS
    196 mm
    1 * 2 ST
    1 * 2 ST - 68,60 €
    68,60 €
    1 * 2 ST
       68,60 €
  • 2000 ml
    24/29 NS
    215 mm
    1 * 1 ST
    1 * 1 ST - 56,70 €
    56,70 €
    1 * 1 ST
       56,70 €
  • 2000 ml
    29/32 NS
    220 mm
    1 * 1 ST
    1 * 1 ST - 56,70 €
    56,70 €
    1 * 1 ST
       56,70 €
  • 2000 ml
    34/35 NS
    228 mm
    1 * 1 ST
    1 * 1 ST - 49,50 €
    49,50 €
    1 * 1 ST
       49,50 €
  • 3000 ml
    24/29 NS
    236 mm
    1 * 5 ST
    1 * 5 ST - 561,00 €
    561,00 €
    1 * 5 ST
       561,00 €
  • 3000 ml
    34/35 NS
    249 mm
    1 * 5 ST
    1 * 5 ST - 553,00 €
    553,00 €
    1 * 5 ST
       553,00 €
  • 5000 ml
    29/32 NS
    270 mm
    1 * 1 ST
    1 * 1 ST - 197,00 €
    197,00 €
    1 * 1 ST
       197,00 €
  • 5000 ml
    34/35 NS
    280 mm
    1 * 1 ST
    1 * 1 ST - 179,00 €
    179,00 €
    1 * 1 ST
       179,00 €