Orders LinkContactUsLinkComponent
Flasks, round bottom, with standard ground joint
Flasks, round bottom, with standard ground joint
Supplier:   Lenz Laborglas
Flasks, round bottom, with standard ground joint
Supplier:  Lenz Laborglas
Certificates
Flasks, round bottom, with standard ground joint
Flasks, round bottom, with standard ground joint
40 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
DIN 12348
Certifications: DIN EN ISO 4797
Documents
Product Family Options
Product Information
  • Capacity
    Socket size
    Height
    Ext.Ø
    Availability
    Price
  • 5 ml
    14/23 NS
    58 ± 3 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,90 €
    9,90 €
    1 * 1 ST
       9,90 €
  • 10 ml
    14/23 NS
    70 ± 3 mm
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,90 €
    8,90 €
    1 * 1 ST
       8,90 €
  • 25 ml
    14/23 NS
    85 ± 3 mm
    41 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,10 €
    11,10 €
    1 * 1 ST
       11,10 €
  • 25 ml
    19/26 NS
    85 ± 3 mm
    41 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,70 €
    9,70 €
    1 * 1 ST
       9,70 €
  • 50 ml
    14/23 NS
    90 ± 6 mm
    51 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,80 €
    11,80 €
    1 * 1 ST
       11,80 €
  • 50 ml
    19/26 NS
    90 ± 6 mm
    51 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,80 €
    10,80 €
    1 * 1 ST
       10,80 €
  • 50 ml
    24/29 NS
    90 ± 6 mm
    51 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,40 €
    13,40 €
    1 * 1 ST
       13,40 €
  • 50 ml
    29/32 NS
    90 ± 6 mm
    51 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,30 €
    14,30 €
    1 * 1 ST
       14,30 €
  • 100 ml
    14/23 NS
    105 ± 6 mm
    64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,20 €
    13,20 €
    1 * 1 ST
       13,20 €
  • 100 ml
    19/26 NS
    105 ± 6 mm
    64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,30 €
    12,30 €
    1 * 1 ST
       12,30 €
  • 100 ml
    24/29 NS
    105 ± 6 mm
    64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,50 €
    13,50 €
    1 * 1 ST
       13,50 €
  • 100 ml
    29/32 NS
    105 ± 6 mm
    64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,20 €
    12,20 €
    1 * 1 ST
       12,20 €
  • 250 ml
    14/23 NS
    140 ± 6 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,10 €
    28,10 €
    1 * 1 ST
       28,10 €
  • 250 ml
    19/26 NS
    140 ± 6 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,00 €
    47,00 €
    1 * 1 ST
       47,00 €
  • 250 ml
    24/29 NS
    140 ± 6 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,00 €
    24,00 €
    1 * 1 ST
       24,00 €
  • 250 ml
    29/32 NS
    140 ± 6 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,70 €
    22,70 €
    1 * 1 ST
       22,70 €
  • 250 ml
    45/40 NS
    140 ± 6 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,90 €
    56,90 €
    1 * 1 ST
       56,90 €
  • 500 ml
    19/26 NS
    163 ± 6 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,80 €
    34,80 €
    1 * 1 ST
       34,80 €
  • 500 ml
    24/29 NS
    163 ± 6 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,70 €
    30,70 €
    1 * 1 ST
       30,70 €
  • 500 ml
    29/32 NS
    163 ± 6 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,00 €
    29,00 €
    1 * 1 ST
       29,00 €
  • 500 ml
    45/40 NS
    163 ± 6 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,30 €
    63,30 €
    1 * 1 ST
       63,30 €
  • 1000 ml
    24/29 NS
    200 ± 7 mm
    131 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,10 €
    44,10 €
    1 * 1 ST
       44,10 €
  • 1000 ml
    29/32 NS
    200 ± 7 mm
    131 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,10 €
    42,10 €
    1 * 1 ST
       42,10 €
  • 1000 ml
    34/35 NS
    200 ± 7 mm
    131 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 74,20 €
    74,20 €
    1 * 1 ST
       74,20 €
  • 1000 ml
    45/40 NS
    200 ± 7 mm
    131 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,20 €
    53,20 €
    1 * 1 ST
       53,20 €
  • 2000 ml
    24/29 NS
    240 ± 7 mm
    166 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 81,30 €
    81,30 €
    1 * 1 ST
       81,30 €
  • 2000 ml
    45/40 NS
    240 ± 7 mm
    166 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 81,50 €
    81,50 €
    1 * 1 ST
       81,50 €
  • 2000 ml
    29/32 NS
    240 ± 7 mm
    166 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,10 €
    72,10 €
    1 * 1 ST
       72,10 €
  • 3000 ml
    29/32 NS
    265 ± 7 mm
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 162,00 €
    162,00 €
    1 * 1 ST
       162,00 €
  • 3000 ml
    45/40 NS
    265 ± 7 mm
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 169,00 €
    169,00 €
    1 * 1 ST
       169,00 €
  • 4000 ml
    29/32 NS
    290 ± 7 mm
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 210,00 €
    210,00 €
    1 * 1 ST
       210,00 €
  • 4000 ml
    45/40 NS
    290 ± 7 mm
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
  • 5000 ml
    29/32 NS
    315 ± 7 mm
    223 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 259,00 €
    259,00 €
    1 * 1 ST
       259,00 €
  • 5000 ml
    45/40 NS
    315 ± 7 mm
    223 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 252,00 €
    252,00 €
    1 * 1 ST
       252,00 €
  • 6000 ml
    29/32 NS
    360 ± 7 mm
    236 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 270,00 €
    270,00 €
    1 * 1 ST
       270,00 €
  • 6000 ml
    45/40 NS
    360 ± 7 mm
    223 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 280,00 €
    280,00 €
    1 * 1 ST
       280,00 €
  • 10000 ml
    29/32 NS
    375 ± 7 mm
    279 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 550,00 €
    550,00 €
    1 * 1 ST
       550,00 €
  • 10000 ml
    45/40 NS
    420 ± 7 mm
    279 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 485,00 €
    485,00 €
    1 * 1 ST
       485,00 €
  • 20000 ml
    29/32 NS
    495 ± 7 mm
    345 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 766,00 €
    766,00 €
    1 * 1 ST
       766,00 €
  • 20000 ml
    45/40 NS
    495 ± 7 mm
    345 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 809,00 €
    809,00 €
    1 * 1 ST
       809,00 €
Cork rings for round bottom flasks
Product Information
  • Catalog #
    Ext.Ø
    For
    Availability
    Price
  • 80 mm
    10 - 100 ml flasks
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 40,30 €
    40,30 €
    1 * 5 ST
       40,30 €
Product Information
  • Catalog #
    Ext.Ø
    For
    Availability
    Price
  • 140 mm
    500 - 1000 ml flasks
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 41,50 €
    41,50 €
    1 * 3 ST
       41,50 €
Product Information
  • Catalog #
    Ext.Ø
    For
    Availability
    Price
  • 170 mm
    2000 ml flasks
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 53,30 €
    53,30 €
    1 * 3 ST
       53,30 €
Product Information
  • Catalog #
    Ext.Ø
    For
    Availability
    Price
  • 210 mm
    4000 ml flasks
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 70,50 €
    70,50 €
    1 * 2 ST
       70,50 €
Product Information
  • Catalog #
    Ext.Ø
    For
    Availability
    Price
  • 240 mm
    6000 ml flasks
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 91,50 €
    91,50 €
    1 * 2 ST
       91,50 €
Product Information
  • Catalog #
    Ext.Ø
    For
    Availability
    Price
  • 110 mm
    250 - 500 ml flasks
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 44,80 €
    44,80 €
    1 * 5 ST
       44,80 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.