Flasks, flat bottom with Quickfit® ground joint
Supplier: SciLabware
Jump to:
About this item
PYREX®, borosilicate glass, clear, with short neck.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat base allows use with hotplates. Labelling area.
22 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, clear, with short neck.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat base allows use with hotplates. Labelling area.
Product Family Options
Product Information
- CapacityHeightSocket sizeAvailabilityPrice