Orders LinkContactUsLinkComponent
Flasks, Erlenmeyer, with Quickfit® ground joint
Flasks, Erlenmeyer, with Quickfit® ground joint
Supplier:   SciLabware
Flasks, Erlenmeyer, with Quickfit® ground joint
Supplier:  SciLabware
Certificates
Flasks, Erlenmeyer, with Quickfit® ground joint
Flasks, Erlenmeyer, with Quickfit® ground joint
22 Options Available Below
Product Details & Documents
PYREX® borosilicate glass, clear. With matt white labelling area.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
ISO 4797
Certificeringen: ISO 4797
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Socket size
    Availability
    Price
  • 25 ml
    70 mm
    42 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 82,90 €
    82,90 €
    1 * 5 ST
       82,90 €
  • 25 ml
    70 mm
    42 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 83,70 €
    83,70 €
    1 * 5 ST
       83,70 €
  • 50 ml
    85 mm
    51 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 94,40 €
    94,40 €
    1 * 5 ST
       94,40 €
  • 50 ml
    85 mm
    51 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 83,10 €
    83,10 €
    1 * 5 ST
       83,10 €
  • 50 ml
    90 mm
    51 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 89,70 €
    89,70 €
    1 * 5 ST
       89,70 €
  • 100 ml
    100 mm
    64 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 74,80 €
    74,80 €
    1 * 5 ST
       74,80 €
  • 100 ml
    100 mm
    64 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 129,00 €
    129,00 €
    1 * 5 ST
       129,00 €
  • 100 ml
    100 mm
    64 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 120,00 €
    120,00 €
    1 * 5 ST
       120,00 €
  • 100 ml
    100 mm
    64 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 63,40 €
    63,40 €
    1 * 5 ST
       63,40 €
  • 150 ml
    118 mm
    74 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 82,40 €
    82,40 €
    1 * 5 ST
       82,40 €
  • 150 ml
    118 mm
    74 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 88,60 €
    88,60 €
    1 * 5 ST
       88,60 €
  • 150 ml
    116 mm
    74 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 85,70 €
    85,70 €
    1 * 5 ST
       85,70 €
  • 250 ml
    130 mm
    85 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 137,00 €
    137,00 €
    1 * 5 ST
       137,00 €
  • 250 ml
    130 mm
    85 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 131,00 €
    131,00 €
    1 * 5 ST
       131,00 €
  • 250 ml
    130 mm
    85 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 81,30 €
    81,30 €
    1 * 5 ST
       81,30 €
  • 250 ml
    130 mm
    85 mm
    34/35 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 143,00 €
    143,00 €
    1 * 5 ST
       143,00 €
  • 500 ml
    175 mm
    101 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 398,00 €
    398,00 €
    1 * 20 ST
       398,00 €
  • 500 ml
    175 mm
    101 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 146,00 €
    146,00 €
    1 * 5 ST
       146,00 €
  • 500 ml
    175 mm
    101 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 95,90 €
    95,90 €
    1 * 5 ST
       95,90 €
  • 1000 ml
    213 mm
    130 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 61,70 €
    61,70 €
    1 * 2 ST
       61,70 €
  • 1000 ml
    220 mm
    130 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 286,00 €
    286,00 €
    1 * 10 ST
       286,00 €
  • 2000 ml
    270 mm
    160 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 288,00 €
    288,00 €
    1 * 5 ST
       288,00 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.