ContactUsLinkComponent
Fernbach culture flasks, Nalgene®
Fernbach culture flasks, Nalgene®
Catalog # 710-4100
   Low Manufacturing Impact
undefined
Fernbach culture flasks, Nalgene®
Catalog # 710-4100
   Low Manufacturing Impact
Specifications
  • Capacity:
    2800 ml
  • Description:
    Fernbach flask
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Specifications
About this item
Clear PC.
  • Thermal resistance from -135 to +135 °C, autoclavable
  • Shatter-proof with high mechanical resistance
  • Good chemical resistance, but not suitable for solvents, strong acids or alkaline solutions
Provides the large liquid surface area necessary for growing microorganisms in liquid culture media. Provides for faster growth through  improved ventilation.