Orders LinkContactUsLinkComponent
Flasks, cylindrical, conical
Flasks, cylindrical, conical
Supplier:   SciLabware
Flasks, cylindrical, conical
Supplier:  SciLabware
Certificates
Flasks, cylindrical, conical
Flasks, cylindrical, conical
3 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • Good thermal resistance
  • With labelling area
Ideal for fermentation and culture applications.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Neck Ø ext.
    Availability
    Price
  • 60 ml
    70 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 1 040,00 €
    1 040,00 €
    1 * 50 ST
       1 040,00 €
  • 125 ml
    97 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,90 €
    22,90 €
    1 * 1 ST
       22,90 €
  • 250 ml
    130 mm
    34 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 1 490,00 €
    1 490,00 €
    1 * 50 ST
       1 490,00 €