Erlenmeyer flasks with thread, DURAN®
Supplier: DWK Life Sciences
Jump to:
About this item
DURAN®, borosilicate glass 3.3, clear, without or with red PBT screw cap.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. A PTFE-coated seal incorporated in the cap guarantees that the flasks are absolutely leakproof.
With printed Retrace Code (batch identification).
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, without or with red PBT screw cap.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. A PTFE-coated seal incorporated in the cap guarantees that the flasks are absolutely leakproof.
With printed Retrace Code (batch identification).
Documents
Product Family Options
With thread and screw cap