ContactUsLinkComponent
Caps for cell culture flasks, Nunc™ Cell Factory™
Caps for cell culture flasks, Nunc™ Cell Factory™
Catalog # 734-2152
Caps for cell culture flasks, Nunc™ Cell Factory™
Catalog # 734-2152
Specifications
  • Description:
    Easy fill vent/close caps for cell factory systems (all models)
  • Pk:
    100
Specifications
About this item
These caps reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the cell factory system.
Further the development of applications requires a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ accessories.