Orders LinkContactUsLinkComponent
First-aid kits
First-aid kits
First-aid kits
Certificates
First-aid kits
7 Options Available Below
Product Details & Documents
The contents of the following first aid kits meet the requirements as specified in the HSE Approved Code of Practice. The code of practice states 'An employer should make an assessment of the first aid needs appropriate to the circumstance of each workplace'. To assist in selecting the appropriate kit, employee ratios are given as guidance provided by the British Healthcare Trade Association.
Delivery information: All kits are supplied with a first aid guidance leaflet.
Specifications for Product Family
Specifications
  • Catalog No:
  • 118-0122
  • 118-0130
  • 118-0132
  • 118-0134
  • 118-0135
  • 118-0121
  • 118-0129
  • Model:
  • Standard (11 - 20 persons)
  • Standard (21 - 50 persons)
  • Industrial (1 - 20 persons)
  • Industrial (1 - 50 persons)
  • Contents:
  • 40 × Waterproof plasters, sterile
    10 × individually wrapped wipes
    4 × eye pads with bandages, non woven
    6 × triangular bandages, non woven
    12 × Safety pins
    9 × sterile dressing, medium size
    3 × sterile dressing, large size
    2 pairs of disposable examination gloves
    1 wall bracket
  • 60 × Waterproof plasters, sterile
    20 × individually wrapped wipes
    6 × eye pads with bandages, non woven
    8 × triangular bandages, non woven
    12 × Safety pins
    12 × sterile dressing, medium size
    4 × sterile dressing, large size
    3 pairs of disposable examination gloves
    1 wall bracket
  • 120 × Waterproof plasters, sterile
    40 × individually wrapped wipes
    12 × blue plastic fingerstalls (assorted)
    4 × eye pads with bandages, non woven
    6 × triangular bandages, non woven
    12 × Safety pins
    9 × sterile dressing, medium size
    3 × sterile dressing, large size
    2 × skin closure adhesives, sterile
    10 × eyewash, sterile
    2 × hygienic resuscitation kit
    8 × Primapore wound dressings (80×80 mm)
    4 × Primapore wound dressings (80×80 mm)
    1 × emergency foil blanket
    1 × dressing scissors
    100 disposable examination gloves (pairs)
    1 wall bracket
  • 120 × Waterproof plasters, sterile
    40 × individually wrapped wipes
    12 × blue plastic fingerstalls (assorted)
    6 × eye pads with bandages, non woven
    8 × triangular bandages, non woven
    12 × Safety pins
    12 × sterile dressing, medium size
    4 × sterile dressing, large size
    2 × skin closure adhesives, sterile
    10 × eyewash, sterile
    2 × hygienic resuscitation kit
    10 × Primapore wound dressings (80×80 mm)
    6 × Primapore wound dressings (80×80 mm)
    1 × emergency foil blanket
    1 × dressing scissors
    100 disposable examination gloves (pairs)
    1 wall bracket
Specifications
Product Family Options
Statutory first aid kits – low risk
Product Information
  • Availability
    Price
  • 118-0122
    Medium first aid kit, 11 - 20 people
    Medium first aid kit, 11 - 20 people
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 135,00 €
    135,00 €
    1 * 1 KIT
       135,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    118-0122
    Model
    Standard (11 - 20 persons)
    Contents
    40 × Waterproof plasters, sterile
    10 × individually wrapped wipes
    4 × eye pads with bandages, non woven
    6 × triangular bandages, non woven
    12 × Safety pins
    9 × sterile dressing, medium size
    3 × sterile dressing, large size
    2 pairs of disposable examination gloves
    1 wall bracket
  • 118-0130
    Large first aid kit, 21 - 50 people
    Large first aid kit, 21 - 50 people
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 113,00 €
    113,00 €
    1 * 1 KIT
       113,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    118-0130
    Model
    Standard (21 - 50 persons)
    Contents
    60 × Waterproof plasters, sterile
    20 × individually wrapped wipes
    6 × eye pads with bandages, non woven
    8 × triangular bandages, non woven
    12 × Safety pins
    12 × sterile dressing, medium size
    4 × sterile dressing, large size
    3 pairs of disposable examination gloves
    1 wall bracket
Refills for first aid kits – low risk
Product Information
  • Availability
    Price
  • 118-0132
    Refill for standard kit
    Refill for standard kit
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 16,50 €
    16,50 €
    1 * 1 KIT
       16,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    118-0132
    Model
    Contents
  • 118-0134
    Refill for medium kit
    Refill for medium kit
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 57,10 €
    57,10 €
    1 * 1 KIT
       57,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    118-0134
    Model
    Contents
  • 118-0135
    Refill for large kit
    Refill for large kit
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 45,10 €
    45,10 €
    1 * 1 KIT
       45,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    118-0135
    Model
    Contents
Statutory first aid kits – high risk
Product Information
  • Availability
    Price
  • 118-0121
    Industrial first aid kit, 1 - 20 people, supplied in shatterproof ABS case
    Industrial first aid kit, 1 - 20 people, supplied in shatterproof ABS case
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 292,00 €
    292,00 €
    1 * 1 KIT
       292,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    118-0121
    Model
    Industrial (1 - 20 persons)
    Contents
    120 × Waterproof plasters, sterile
    40 × individually wrapped wipes
    12 × blue plastic fingerstalls (assorted)
    4 × eye pads with bandages, non woven
    6 × triangular bandages, non woven
    12 × Safety pins
    9 × sterile dressing, medium size
    3 × sterile dressing, large size
    2 × skin closure adhesives, sterile
    10 × eyewash, sterile
    2 × hygienic resuscitation kit
    8 × Primapore wound dressings (80×80 mm)
    4 × Primapore wound dressings (80×80 mm)
    1 × emergency foil blanket
    1 × dressing scissors
    100 disposable examination gloves (pairs)
    1 wall bracket
  • 118-0129
    Industrial first aid kit, 1 - 50 people, supplied in shatterproof ABS case
    Industrial first aid kit, 1 - 50 people, supplied in shatterproof ABS case
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 648,00 €
    648,00 €
    1 * 1 KIT
       648,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    118-0129
    Model
    Industrial (1 - 50 persons)
    Contents
    120 × Waterproof plasters, sterile
    40 × individually wrapped wipes
    12 × blue plastic fingerstalls (assorted)
    6 × eye pads with bandages, non woven
    8 × triangular bandages, non woven
    12 × Safety pins
    12 × sterile dressing, medium size
    4 × sterile dressing, large size
    2 × skin closure adhesives, sterile
    10 × eyewash, sterile
    2 × hygienic resuscitation kit
    10 × Primapore wound dressings (80×80 mm)
    6 × Primapore wound dressings (80×80 mm)
    1 × emergency foil blanket
    1 × dressing scissors
    100 disposable examination gloves (pairs)
    1 wall bracket