Order Entry
Belgium
ContactUsLinkComponent
 
Venting filters, Vacushield™
  514-4115
undefined
Venting filters, Vacushield™
  514-4115

 

  • Ø:
    50 mm
  • Filtration area:
    19,6 cm²
  • Pore size:
    0,2 µm
  • Max. temperature:
    130 °C
  • Seq:
    01
  • Inlet/outlet:
    For tubing Ø 6,4 - 12,7
  • Max. pressure:
    1 bar
  • Airflow rate:
    8 L/min (0,2 bar)
  • Pk:
    3

 

 

Protects valves and pump components from damage due to liquids. Protects laboratory personnel from potential biohazards, airborne and aerosol contaminants.

Hydrophobic PTFE membrane, PP housing

Aerosol retention: 99,97% of 0,3 µm DOP particles at 32 L/min/100 cm²