ContactUsLinkComponent
Vent filters, HEPA-CAP, Whatman™
Vent filters, HEPA-CAP, Whatman™
Certificates
undefined
Vent filters, HEPA-CAP, Whatman™
Certificates
Specifications
  • Catalog No:
  • 516-1104
  • 516-1105
  • 517-0011
  • 517-0008
  • 517-0012
  • Ø:
  • 36 mm
  • 75 mm
  • 75 mm
  • 36 mm
  • 150 mm
  • Housing type:
  • Hepa-Cap™ 36 capsule
  • Hepa-Cap™ 75 capsule
  • Hepa-Cap™ 75 capsule
  • Hepa-Cap™ 36 capsule
  • Hepa-Cap™ 150 capsule
  • Airflow rate:
  • 225 L/min (0,35 bar)
  • 700 L/min (0,35 bar)
  • 225 L/min (0,35 bar)
  • 225 L/min (0,35 bar)
  • 225 L/min (0,35 bar)
  • Max. pressure:
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • Inlet/outlet:
  • 1/4 to 3/8" stepped barb
  • 3/8 to 1/2" stepped barb
  • 3/8" FNPT
  • 3/8" FNPT
  • 3/8" FNPT