Orders LinkContactUsLinkComponent
Membrane filters, Microsart® e.motion
Membrane filters, Microsart® e.motion
Supplier:  Sartorius
Certificates
Membrane filters, Microsart® e.motion
Membrane filters, Microsart® e.motion
23 Options Available Below
Product Details & Documents
These sterile cellulose nitrate and PES Microsart® e.motion membrane filters can be used for colony counting, particle testing and microscopy. They are specifically designed for use in the Microsart® e.motion dispenser.
  • Outstanding recovery rates for microorganisms
  • Product data is printed on individual envelopes
  • Gamma irradiated, 25 kGray
The special pleating of the band of membrane filters ensures that they are perfectly flat when dispensed. The membrane filters can also be used with nutrient pad sets.
 : 0,45 µm filters are according to ISO 7704
 : Individually sterile sealed in a format packaged on a pleated band, without protective paper.
Product Family Options
Product Information
  • Pore size
    Ø
    Availability
    Price
  • 514-7019
    White with black grid, cellulose nitrate
    White with black grid, cellulose nitrate
    0,20 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 515-0241
    White without grid, PES
    White without grid, PES
    0,20 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 433,00 €
    433,00 €
    1 * 300 ST
       433,00 €
  • 514-7020
    White with black grid, cellulose nitrate
    White with black grid, cellulose nitrate
    0,20 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-0057
    White with green grid, high flow, cellulose nitrate
    White with green grid, high flow, cellulose nitrate
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-0076
    White with black grid, cellulose nitrate
    White with black grid, cellulose nitrate
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-0077
    Grey with white grid, cellulose nitrate
    Grey with white grid, cellulose nitrate
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-0078
    Green with dark green grid, cellulose nitrate
    Green with dark green grid, cellulose nitrate
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-0079
    White with green grid, cellulose nitrate
    White with green grid, cellulose nitrate
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-7015
    White with black grid, high flow, cellulose nitrate
    White with black grid, high flow, cellulose nitrate
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • SART114H6Z-47-SFM
    White with black grid, high flow, cellulose nitrate
    White with black grid, high flow, cellulose nitrate
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 450 ST
    1 * 450 ST - 511,00 €
    511,00 €
    1 * 450 ST
       511,00 €
  • SART114H6Z-47-SJR
    White with black grid, high flow, cellulose nitrate, big pack
    White with black grid, high flow, cellulose nitrate, big pack
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 728,00 €
    728,00 €
    1 * 1.000 ST
       728,00 €
  • 515-0231
    Grey with white grid, high flow, cellulose nitrate
    Grey with white grid, high flow, cellulose nitrate
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • SART130H6Z-47-SJR
    Grey with white grid, high flow, cellulose nitrate, big pack
    Grey with white grid, high flow, cellulose nitrate, big pack
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 733,00 €
    733,00 €
    1 * 1 ST
       733,00 €
  • 514-7018
    White with black grid, cellulose nitrate
    White with black grid, cellulose nitrate
    0,45 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-7024
    Grey with white grid, cellulose nitrate
    Grey with white grid, cellulose nitrate
    0,45 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-7025
    Green with dark green grid, cellulose nitrate
    Green with dark green grid, cellulose nitrate
    0,45 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-7026
    White with green grid, cellulose nitrate
    White with green grid, cellulose nitrate
    0,45 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • SART114H6Z-50-SCM
    White with black grid, cellulose nitrate
    White with black grid, cellulose nitrate
    0,45 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • SART130H6Z-50-SCM
    Grey with white grid, high flow, cellulose nitrate
    Grey with white grid, high flow, cellulose nitrate
    0,45 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-0058
    Grey with white grid, cellulose nitrate
    Grey with white grid, cellulose nitrate
    0,65 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-7023
    Grey with white grid, cellulose nitrate
    Grey with white grid, cellulose nitrate
    0,65 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-7021
    Grey with white grid, cellulose nitrate
    Grey with white grid, cellulose nitrate
    0,80 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €
  • 514-7016
    White with black grid, cellulose nitrate
    White with black grid, cellulose nitrate
    1,20 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 407,00 €
    407,00 €
    1 * 300 ST
       407,00 €