Glass microfibre filters with binder
These filters are mostly used either for monitoring air and gas or as prefilter. They are manufactured with synthetic binding agents to ensure that the filter has a defined strength.
- Mechanically and chemically stable
- Hydrophilic
- Catalog No:
- SARTFT-3-01125-110
- SART13400-25-Q
- SART13400-16-S
- SARTFT-3-01125-130
- SARTFT-3-01125-125
- 512-0034
- 512-0051
- 512-0035
- SARTFT-3-01125-045
- SARTFT-3-01125-050
- SART13400-80-N
- 512-0053
- 512-0054
- 512-0036
- SARTFT-3-01125-047
- 512-0064
- 512-0055
- SART13430-100-K
- 512-0033
- SART13400-100-K
- SART13400-13-S
- SART13400-20-S
- 512-0065
- Grade:
- MG 1387/1
- 13400
- 13400
- MG 1387/1
- MG 1387/1
- 13400
- 13400
- 13400
- MG 1387/1
- MG 1387/1
- 13400
- 13400
- 13400
- 13400
- MG 1387/1
- 13430
- 13400
- 13430
- 13400
- 13400
- 13400
- 13400
- 13430
- Weight:
- 90 g/m²
- 74 g/m²
- 74 g/m²
- 90 g/m²
- 90 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 90 g/m²
- 90 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 90 g/m²
- 223 g/m²
- 74 g/m²
- 223 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 223 g/m²
- Thickness:
- 0,40 mm
- 0,34 mm
- 0,34 mm
- 0,40 mm
- 0,40 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,40 mm
- 0,40 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,40 mm
- 1,30 mm
- 0,34 mm
- 1,30 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 1,30 mm
- Max. temperature:
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
Frequently Bought Together