Orders LinkContactUsLinkComponent
Quantitative filter papers
   Sustainable Options Available
Supplier:   Sartorius
Quantitative filter papers
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Quantitative filter papers
16 Options Available Below
Product Details & Documents
These filter papers are made of 100% cotton linters. They are guaranteed to be free of any residual acids and they have an extremely low percentage of ash content. As a result, these filters are virtually ash-free. All Sartorius ashless filter papers are characterised by high wet strength and high purity and are therefore especially suitable for quantitative analysis, Büchner funnels, pressure or vacuum filtration.
388: 'Black dot filter' For coarse-flocculent and voluminous precipitates, especially iron, aluminium and chromium hydroxides; copper, bismuth, cobalt and iron sulphides; various organic, coloured metal deposits; determination of silicon in analysis of steel and pig iron; for acidic and slightly alkaline solutions; pressure filtration.
389: 'White dot filter' The most used grade for analyses; for relatively coarse precipitates such as silver, arsenic, antimony, cadmium, lead, iron and manganese sulphides; lead chromate; ammonium phosphorous molybdate; alkaline earth carbonates '...' for acidic and slightly alkaline solutions; pressure filtration.
390: 'Green dot filter' Filtration of fine precipitates; barium sulphate; lead molybdate; lead dioxide; calcium hydroxide; calcium fluoride; nickel, zinc and stannic sulphides; for acidic and slightly alkaline solutions; pressure filtration.
393: 'Purple dot filter' Ideal for particularly difficult filtration conditions and especially fine-grained precipitates; barium sulphate; copper oxide.
Specifications for Product Family
Specifications
  • Catalog No:
  • 515-1202
  • 515-1203
  • 516-7042
  • 515-1162
  • 515-1143
  • 515-1183
  • 516-7044
  • 515-1184
  • 515-1226
  • 515-1146
  • 515-1208
  • SARTFT-3-101-400
  • SARTFT-2-101580580
  • SARTFT-2-102580580
  • SARTFT-2-103580580
  • SARTFT-2-127580580
  • Grade:
  • 391
  • 391
  • 393
  • 389
  • 388
  • 390
  • 393
  • 390
  • 392
  • 388
  • 391
  • 388
  • 388
  • 389
  • 390
  • 393
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Weight:
  • 84 g/m²
  • 84 g/m²
  • 100 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 100 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 100 g/m²
  • Particle retention:
  • 2 - 3 µm
  • 2 - 3 µm
  • 1 - 2 µm
  • 8 - 12 µm
  • 12 - 15 µm
  • 3 - 5 µm
  • 1 - 2 µm
  • 3 - 5 µm
  • 5 - 8 µm
  • 12 - 15 µm
  • 2 - 3 µm
  • 12 - 15 µm
  • 12 - 15 µm
  • 8 - 12 µm
  • 3 - 5 µm
  • 1 µm
  • Filtration speed:
  • 180 s
  • 180 s
  • 300 s
  • 20 s
  • 10 s
  • 100 s
  • 300 s
  • 100 s
  • 50 s
  • 10 s
  • 180 s
  • 10 s
  • 10 s
  • 20 s
  • 100 s
  • 300 s
  • Ash content:
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • ≤0,010%
  • Environmental certification:
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
Specifications
Product Family Options
Product Information
  • Grade
    Size
    Availability
    Price
  • 391
    55 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 13,40 €
    13,40 €
    1 * 100 ST
       13,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1202
    Grade
    391
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    2 - 3 µm
    Filtration speed
    180 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 391
    70 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 17,50 €
    17,50 €
    1 * 100 ST
       17,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1203
    Grade
    391
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    2 - 3 µm
    Filtration speed
    180 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 393
    70 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 17,50 €
    17,50 €
    1 * 100 ST
       17,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    516-7042
    Grade
    393
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    100 g/m²
    Particle retention
    1 - 2 µm
    Filtration speed
    300 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 389
    70 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 17,50 €
    17,50 €
    1 * 100 ST
       17,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1162
    Grade
    389
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    8 - 12 µm
    Filtration speed
    20 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 388
    90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 28,10 €
    28,10 €
    1 * 100 ST
       28,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1143
    Grade
    388
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    12 - 15 µm
    Filtration speed
    10 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 390
    90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 28,10 €
    28,10 €
    1 * 100 ST
       28,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1183
    Grade
    390
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    3 - 5 µm
    Filtration speed
    100 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 393
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 38,70 €
    38,70 €
    1 * 100 ST
       38,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    516-7044
    Grade
    393
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    100 g/m²
    Particle retention
    1 - 2 µm
    Filtration speed
    300 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 390
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 38,70 €
    38,70 €
    1 * 100 ST
       38,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1184
    Grade
    390
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    3 - 5 µm
    Filtration speed
    100 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 392
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 45,60 €
    45,60 €
    1 * 100 ST
       45,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1226
    Grade
    392
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    5 - 8 µm
    Filtration speed
    50 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 388
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 65,10 €
    65,10 €
    1 * 100 ST
       65,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1146
    Grade
    388
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    12 - 15 µm
    Filtration speed
    10 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 391
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 97,90 €
    97,90 €
    1 * 100 ST
       97,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1208
    Grade
    391
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    2 - 3 µm
    Filtration speed
    180 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 388
    400 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 366,00 €
    366,00 €
    1 * 100 ST
       366,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SARTFT-3-101-400
    Grade
    388
    Environmentally Preferable
    Weight
    84 g/m²
    Particle retention
    12 - 15 µm
    Filtration speed
    10 s
    Ash content
    ≤0,010%
    Environmental certification
  • 388
    580×580 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 782,00 €
    782,00 €
    1 * 100 ST
       782,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-2-101580580
    Grade
    388
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    12 - 15 µm
    Filtration speed
    10 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 389
    580×580 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 782,00 €
    782,00 €
    1 * 100 ST
       782,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-2-102580580
    Grade
    389
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    8 - 12 µm
    Filtration speed
    20 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 390
    580×580 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 782,00 €
    782,00 €
    1 * 100 ST
       782,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-2-103580580
    Grade
    390
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    84 g/m²
    Particle retention
    3 - 5 µm
    Filtration speed
    100 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 393
    580×580 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 782,00 €
    782,00 €
    1 * 100 ST
       782,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-2-127580580
    Grade
    393
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    100 g/m²
    Particle retention
    1 µm
    Filtration speed
    300 s
    Ash content
    ≤0,010%
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact