Orders LinkContactUsLinkComponent
Phase separating paper, grade 1PS
Phase separating paper, grade 1PS
Certificates
Phase separating paper, grade 1PS
Phase separating paper, grade 1PS
8 Options Available Below
Product Details & Documents
1PS phase-separating paper is a hydrophobic filter for routine solvent extraction. Within seconds it retains aqueous phases, while solvent phases flow through.
  • Impregnated with stabilised silicone to create a hydrophobic filter
  • Multiple separations can be processed in parallel
  • No special training required
  • Can replace a separatory funnel in certain applications
Solvent extraction can be time-consuming and messy when numerous separations are required
Mixed phases are poured directly into the 1PS phase-separating paper. The solvent phase flows through the paper quickly and cleanly, automatically stopping after the process is complete; all of the aqueous phase is retained. Droplets are automatically separated within seconds.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 70 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 54,80 €
    54,80 €
    1 * 100 ST
       54,80 €
  • 90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 75,80 €
    75,80 €
    1 * 100 ST
       75,80 €
  • 110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 75,20 €
    75,20 €
    1 * 100 ST
       75,20 €
  • 125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 83,80 €
    83,80 €
    1 * 100 ST
       83,80 €
  • 150 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 152,00 €
    152,00 €
    1 * 100 ST
       152,00 €
  • 185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 139,00 €
    139,00 €
    1 * 100 ST
       139,00 €
  • 240 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 195,00 €
    195,00 €
    1 * 100 ST
       195,00 €
  • 270 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 295,00 €
    295,00 €
    1 * 100 ST
       295,00 €
  • 40 mm × 50 m
    Discontinued