Bottle top vacuum filtration systems, cellulose nitrate membrane, Rapid-Flow™ Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Rapid-Flow™ disposable filter units with cellulose nitrate membrane, PS funnel and collection reservoir feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput.
Color coded according to membrane
Membrane with excellent wetting properties and fastest flow rates with aqueous solutions
Funnel and receiver flask graduated
- Quick disconnect tubing adapter with cover
Sterile disposable filters for cold sterilisation, clarification, prefiltration and ultrafiltration of buffers, culture media and aqueous solutions.
Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit. Available with Fit for Purpose Sterile documentation.
Specifications
- Catalog No:
- 734-5082
- 734-5083
- 734-5084
- 734-5010
- 734-5011
- 734-5008
- 734-5009
- 734-5002
- 734-5006
- 734-5001
- 734-5007
- 734-5004
- 734-5003
- 734-5005
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement