Order Entry
Belgium
ContactUsLinkComponent
Acrodisc® units with Mustang® E membrane
Acrodisc® units with Mustang® E membrane
Catalog # 514-4235
undefined
Acrodisc® units with Mustang® E membrane
Catalog # 514-4235

Specifications

  • Ø:
    25 mm
  • Housing material:
    PP
  • Membrane:
    Mustang® E
  • Pore size:
    0,20 µm
  • Sterility:
    Sterile
  • Max. operating pressure:
    5,5 bar
  • Water flow rate:
    1 - 4 ml/min
  • Pk:
    10
  • Inlet/outlet connections:
    Female Luer lock/male Luer

Specifications

About this item

Remove endotoxin from water, buffer, neutral sugars and certain biological solutions. Also for plasmid preparation for transfection.

Sterilised by gamma irradiation and individually packaged.

Flow rate: 1 to 4 ml/min