ContactUsLinkComponent
Needle holder for specimen loops and dissecting needles
Needle holder for specimen loops and dissecting needles
Catalog # 631-7142
Supplier:  HAMMACHER KARL
undefined
Needle holder for specimen loops and dissecting needles
Catalog # 631-7142
Supplier:  HAMMACHER KARL
Specifications
  • Description:
    Needle holder
  • Length:
    170 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Needle holder for for specimen loops and dissecting needles up to max. diameter of 1,0 mm.