ContactUsLinkComponent
Petri dish racks, Nalgene®
Petri dish racks, Nalgene®
Catalog # 391-2161
   Low Manufacturing Impact
undefined
Petri dish racks, Nalgene®
Catalog # 391-2161
   Low Manufacturing Impact
Specifications
  • Description:
    Petri dish rack
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Specifications
About this item
Petri dish rack with PC frame and white PC posts.
  • Smooth, rounded corners
  • Finger-grip handle and rubber feet make carrying safe and secure
  • Rack can be decontaminated by heat sterilisation
For up to 54 plastic round dishes (100×15 mm) or 42 glass round dishes (100×15 mm). Simple assembly required.
W×D×H: 203×343×235 mm