Orders LinkContactUsLinkComponent
Storage pots
Supplier:   KARTELL
Storage pots
Supplier:  KARTELL
Certificates
Storage pots
2 Options Available Below
Product Details & Documents
PP, white.
  • Very good chemical and temperature resistance
  • Robust
  • Impermeable to liquids
Ideal for keeping instruments, pipettes, thermometers and many other laboratory tools tidy and close to hand. With extra wide base for stability.
Product Family Options
Product Information
  • Capacity
    Outer height
    I-height
    Ext.Ø
    Ø int.
    Availability
    Price
  • 230 ml
    106,5 mm
    100 mm
    98 mm
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,30 €
    20,30 €
    1 * 1 ST
       20,30 €
  • 1100 ml
    199,7 mm
    190 mm
    147 mm
    89 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,50 €
    26,50 €
    1 * 1 ST
       26,50 €