Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
Supplier:   Anicrin
Jars with screw cap
Supplier:  Anicrin
Certificates
Jars with screw cap
Jars with screw cap
8 Options Available Below
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
  • Twist cap for quick opening and closing
  • Very good temperature resistance
  • Very good chemical resistance
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Type
    Capacity
    Availability
    Price
  • 150 ml
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 82,10 €
    82,10 €
    1 * 250 ST
       82,10 €
  • with polypropylene screw cap, labelled
    150 ml
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 94,10 €
    94,10 €
    1 * 250 ST
       94,10 €
  • with polypropylene screw cap, irradiated
    150 ml
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 114,00 €
    114,00 €
    1 * 250 ST
       114,00 €
  • with polypropylene screw cap, labelled, irradiated
    150 ml
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 115,00 €
    115,00 €
    1 * 250 ST
       115,00 €
  • with polypropylene screw cap, individually wrapped
    150 ml
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 94,30 €
    94,30 €
    1 * 250 ST
       94,30 €
  • with polypropylene screw cap, irradiated, individually wrapped
    150 ml
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 107,00 €
    107,00 €
    1 * 250 ST
       107,00 €
  • with polypropylene screw cap, individually wrapped, labelled
    150 ml
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 94,30 €
    94,30 €
    1 * 250 ST
       94,30 €
  • with polypropylene screw cap, individually wrapped, labelled, irradiated
    150 ml
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 111,00 €
    111,00 €
    1 * 250 ST
       111,00 €