Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
Supplier:   VITLAB
Jars with screw cap
Supplier:  VITLAB
Certificates
Jars with screw cap
Jars with screw cap
4 Options Available Below
Product Details & Documents
White PP.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Tightly fitting screw lid
To store and transport samples securely.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 30 ml
    38 mm
    54 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 82,00 €
    82,00 €
    1 * 10 ST
       82,00 €
  • 60 ml
    38 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 104,00 €
    104,00 €
    1 * 10 ST
       104,00 €
  • 90 ml
    54 mm
    62 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 128,00 €
    128,00 €
    1 * 10 ST
       128,00 €
  • 180 ml
    54 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 153,00 €
    153,00 €
    1 * 10 ST
       153,00 €