Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
Jars with screw cap
Supplier:  DELTALAB
Certificates
Jars with screw cap
Jars with screw cap
6 Options Available Below
Product Details & Documents
White PP with yellow safety screw cap.
  • Excellent thermal and chemical resistance
  • Graduated
  • Airtight, liquid-tight
Ideal for transporting and storing biological materials, liquids, solutions and powdery materials.
Documents
Product Family Options
Product Information