Orders LinkContactUsLinkComponent
VWR®, Sample Containers, with Attached Lids
VWR®, Sample Containers, with Attached Lids
VWR®, Sample Containers, with Attached Lids
Supplier:  VWR Collection
Certificates
VWR®, Sample Containers, with Attached Lids
VWR®, Sample Containers, with Attached Lids
3 Options Available Below
Product Details & Documents
PP, translucent.
  • Can be used with one hand; the cap remains vertical when open
  • Very good chemical and temperature resistance
  • Aseptic
Ideal for collecting, storing and transporting samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 216-2625
    PP, translucent, hinged lid
    PP, translucent, hinged lid
    50 ml
    31 mm
    78 mm
    Log in for availability
    Log in for availability
    1 * 650 ST
    1 * 650 ST - 173,00 €
    173,00 €
    1 * 650 ST
       173,00 €
  • 90 ml
    43 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 350 ST
    1 * 350 ST - 123,00 €
    123,00 €
    1 * 350 ST
       123,00 €
  • 300 ml
    75 mm
    88 mm
    Log in for availability
    Log in for availability
    1 * 240 ST
    1 * 240 ST - 207,00 €
    207,00 €
    1 * 240 ST
       207,00 €