Seals for threaded connection caps (Septa)
Supplier: BIRZLE PETER
Jump to:
About this item
Silicone, transparent.
- Ideal for use as septa (for puncturing)
- Very good chemical resistance
- Very good thermal resistance
For threaded connection caps with GL thread.
3 Options Available Below
Product Details & Documents
Silicone, transparent.
- Ideal for use as septa (for puncturing)
- Very good chemical resistance
- Very good thermal resistance
For threaded connection caps with GL thread.
Product Family Options
Product Information
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.