Orders LinkContactUsLinkComponent
Air condensers
Air condensers
Supplier:   Lenz Laborglas
Air condensers
Supplier:  Lenz Laborglas
Certificates
Air condensers
2 Options Available Below
Product Details & Documents
DURAN® borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground socket.
Documents
Product Family Options
Product Information
  • Jacket length
    Cone size
    Availability
    Price
  • 1000 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,20 €
    27,20 €
    1 * 1 ST
       27,20 €
  • 700 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,00 €
    19,00 €
    1 * 1 ST
       19,00 €