Anion exchange HPLC columns, RCX-10
Supplier: HAMILTON BONADUZ
Certificates
About this item
RCX-10 carbohydrate analysis columns are designed for the isocratic or gradient separation of carbohydrates. When an isocratic method is used with a conductivity, refractive index, ultraviolet, or pulsed amperometric detector (PAD), mono- and di-saccharides such as glucose, fructose, sucrose and lactose can be quickly determined.
- Greater exchange capacity than that of the PRP-X100
- Samples with two or three carbohydrates can be quickly separated isocratically
- More complex samples require gradient elution
Specifications
- Catalog No:
- HAMI79292
- 554-1254
- 554-1277
- 554-0242
- 554-1276
- 554-1253
- 554-0219
- Max. pressure:
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- pH range:
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- Temperature limit:
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)